Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLN64/STARD3 Mouse anti-Human, Unconjugated, Clone: 1STAR‐2G5, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS1945100UG
Description
MLN64/STARD3 Monoclonal antibody specifically detects MLN64/STARD3 in Human samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blotting.Specifications
MLN64/STARD3 | |
Monoclonal | |
Unconjugated | |
STARD3, CAB1, MLN64 | |
His-tagged recombinant fragment corresponding to 114 amino acids from the START domain of human StAR-related lipid transfer protein 3. | |
Protein G Purified | |
Signaling | |
Clone 1STAR‐2G5 is a mouse monoclonal antibody that detects human StAR-related lipid transfer protein 3 (STARD3). It targets an epitope within the START domain. | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
IgG1 κ |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
1STAR‐2G5 | |
StAR-related lipid transfer protein 3;Metastatic lymph node gene 64 protein;MLN 64;Protein CAB1;START domain-containing protein 3 | |
Mouse | |
50.50 kDa calculated. | |
100 μg | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction