Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLN64/STARD3 Mouse anti-Human, Unconjugated, Clone: 1STAR‐2G5, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen MLN64/STARD3
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABS19451MI
View Documents
MilliporeSigma
MABS1945100UG
100 μg
Each for $372.00
Add to cart
 
MABS19452MI
View Documents
MilliporeSigma
MABS194525UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

MLN64/STARD3 Monoclonal antibody specifically detects MLN64/STARD3 in Human samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blotting.
Specifications

Specifications

MLN64/STARD3
Unconjugated
Mouse
StAR-related lipid transfer protein 3;Metastatic lymph node gene 64 protein;MLN 64;Protein CAB1;START domain-containing protein 3
His-tagged recombinant fragment corresponding to 114 amino acids from the START domain of human StAR-related lipid transfer protein 3.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Immunocytochemistry, Immunoprecipitation, Western Blot
Purified
Human
STARD3, CAB1, MLN64
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit