Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MLNR Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533937
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey (100%) Gibbon (95%) Bovine (89%) Marmoset (84%).
Motilin is a widely conserved 22 amino acid peptide hormone secreted from enterochromaffin cells of the gastrointestinal tract. Within the intestines, motilin potentiates intestinal motility by inducing contractions of the duodenum and through binding to surface receptors, designated GPR38 (G-protein coupled receptor 38). These motilin receptors are predominantly expressed in stomach, thyroid and bone marrow, and they are related to other G-protein coupled receptors located in the pituitary and hypothalamus, which mediate the signaling for growth hormone secretagogues. The gene encoding GPR38 is alternately spliced at the carboxy terminus to generate two related proteins that are designated GPR38-A and GPR38-B. Sequence comparisons of the two isoforms indicate that GPR38-A contains seven transmembrane domains while GPR38-B is predicted to contains only five transmembrane regions. Consistent with other G-protein coupled receptors, GPR38 activates phospholipase C signal transduction pathways and induces intracellular calcium mobilization after binding of motilin.
Specifications
MLNR | |
Polyclonal | |
Unconjugated | |
MLNR | |
G protein-coupled receptor 38; GPR38; G-protein coupled receptor 38; MLNR; Motilin receptor; MTLR; MTLR1; thyrotropin releasing hormone receptor 2; Trhr2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2862 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
O43193 | |
MLNR | |
Synthetic 19 amino acid peptide from internal region of human MLNR. | |
50 μg | |
Primary | |
Human, Monkey | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction