Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melanophilin Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548408

Catalog No. PA548408

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Rab effector protein involved in melanosome transport. Serves as link between melanosome-bound RAB27A and the motor protein MYO5A.
Specifications

Specifications

Melanophilin
Polyclonal
PBS with 0.09% sodium azide
Q9BV36
Melanophilin, Exophilin-3, Slp homolog lacking C2 domains a, SlaC2-a, Synaptotagmin-like protein 2a, MLPH, SLAC2A
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79083
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MLPH
Liquid
MLPH
KLH conjugated synthetic peptide between 541-569 amino acids from the C-terminal region of human MLPH.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit