Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MLSTD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553585
Description
Immunogen sequence: YYEGKNVLLT GATGFLGKVL LEKLLRSCPK VNSVYVLVRQ KAGQTPQERV EEVLSGKLFD RLRDENPDFR EKIIAINSEL TQPKLALSEE DKEVIIDSTN IIFHCAATVR FNENLRDAVQ LNVIATRQLI LLAQQMKNLE VFM Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.
MLSTD2 catalyzes the reduction of saturated fatty acyl-CoA with chain length C16 or C18 to fatty alcohols.
Specifications
MLSTD2 | |
Polyclonal | |
Unconjugated | |
FAR1 | |
2600011M19Rik; 2900034E22Rik; 3732409C05Rik; AI850429; FAR1; fatty acyl CoA reductase 1; fatty acyl-CoA reductase 1; male sterility domain containing 2; male sterility domain-containing protein 2; MLSTD2; PFCRD; SDR10E1; short chain dehydrogenase/reductase family 10E, member 1; UNQ2423/PRO4981 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84188 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WVX9 | |
FAR1 | |
Recombinant protein corresponding to Human MLSTD2. Recombinant protein control fragment (Product #RP-90598). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction