Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMAA Mouse anti-Human, Clone: OTI3A3, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502278

Catalog No. 50-167-3345


Add to cart

Description

Description

The protein encoded by this gene is involved in the translocation of cobalamin into the mitochondrion, where it is used in the final steps of adenosylcobalamin synthesis. Adenosylcobalamin is a coenzyme required for the activity of methylmalonyl-CoA mutase. Defects in this gene are a cause of methylmalonic aciduria.
Specifications

Specifications

MMAA
Monoclonal
0.75 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8IVH4
cblA
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
166785
Immunohistochemistry (Paraffin), Western Blot
OTI3A3
Unconjugated
MMAA
Liquid
MMAA
Full length human recombit protein of human MMAA produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit