Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MMAA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557917
Description
Immunogen sequence: EMWDKMKDFQ DLMLASGELT AKRRKQQKVW MWNLIQESVL EHFRTHPTVR EQIPLLEQKV LIGALSPGLA ADFL Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 74%.
The protein encoded by this gene is involved in the translocation of cobalamin into the mitochondrion, where it is used in the final steps of adenosylcobalamin synthesis. Adenosylcobalamin is a coenzyme required for the activity of methylmalonyl-CoA mutase. Defects in this gene are a cause of methylmalonic aciduria.
Specifications
MMAA | |
Polyclonal | |
Unconjugated | |
MMAA | |
2810018E08Rik; AI840684; cblA; methylmalonic aciduria (cobalamin deficiency) cblA type; methylmalonic aciduria (cobalamin deficiency) type A; methylmalonic aciduria type A homolog, mitochondrial; Methylmalonic aciduria type A protein, mitochondrial; MMAA; mutant adenosylcobalamin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
166785 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IVH4 | |
MMAA | |
Recombinant protein corresponding to Human MMAA. Recombinant protein control fragment (Product #RP-96688). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction