Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMAB Mouse anti-Human, Mouse, Rat, HRP, Clone: OTI2G5, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272730H
Description
MMAB Monoclonal specifically detects MMAB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MMAB | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
MMAB | |
Full length recombinant protein of human MMAB (NP_443077) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2G5 | |
Western Blot, Immunohistochemistry | |
aquocob(I)alamin vitamin B12s adenosyltransferase, ATP:cob(I)alamin adenosyltransferase, ATP:corrinoid adenosyltransferase, ATR, cblB, c-diamide adenosyltransferase, mitochondrial, cob, Cob(I)alamin adenosyltransferase, cob(I)yrinic acid a, EC 2.5.1.17, methylmalonic aciduria (cobalamin deficiency) cblB type, methylmalonic aciduria (cobalamin deficiency) type B, Methylmalonic aciduria type B protein, MGC20496 | |
Mouse | |
Affinity Purified | |
Proteases & Other Enzymes | |
326625.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title