Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMACHC Antibody (OTI4E9), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272734APC
Description
MMACHC Monoclonal specifically detects MMACHC in Human samples. It is validated for Flow Cytometry.Specifications
| MMACHC | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| MMACHC | |
| Full length recombinant protein of human MMACHC (NP_056321) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| OTI4E9 | |
| Flow Cytometry | |
| cblC, DKFZP564I122, FLJ25671, methylmalonic aciduria (cobalamin deficiency) cblC type, with homocystinuria, methylmalonic aciduria and homocystinuria type C protein, RP11-291L19.3 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 25974.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction