Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMACHC Mouse anti-Human, Clone: OTI1A4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526017

Catalog No. PIMA526017


Add to cart

Description

Description

The exact function of the protein encoded by this gene is not known, however, its C-terminal region shows similarity to TonB, a bacterial protein involved in energy transduction for cobalamin (vitamin B12) uptake. Hence, it is postulated that this protein may have a role in the binding and intracellular trafficking of cobalamin. Mutations in this gene are associated with methylmalonic aciduria and homocystinuria type cblC.
Specifications

Specifications

MMACHC
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9Y4U1
RP11-291L19.3, cblC, methylmalonic aciduria and homocystinuria type C protein
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
25974
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1A4
Unconjugated
MMACHC
Liquid
MMACHC
Full length human recombit protein of MMACHC produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit