Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCAM/CD146 Rat anti-Mouse, Alexa Fluor 405, Clone: 733216, R&D Systems
Rat MonoclonalAntibody
Manufacturer: R&D Systems FAB7718V100UG
This item is not returnable.
View return policy
Description
MCAM/CD146 Monoclonal antibody specifically detects MCAM/CD146 in Mouse samples. It is validated for Flow Cytometry.Specifications
MCAM/CD146 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q8R2Y2 | |
MCAM | |
Mouse myeloma cell line NS0-derived recombinant mouse MCAM/CD146 Met1-Val563 Accession # Q8R2Y2 | |
100 ug | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. | |
Monoclonal | |
Detects mouse MCAM/CD146 in direct ELISAs. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse (rm) MAdCAM-1 is observed, and no cross-reactivity with rmALCAM, rmNCAM, rmL1CAM, rmOCAM, rmTROP-2, recombinant human MCAM, or recombinant rat MCAM is observed. |
Flow Cytometry | |
733216 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, L-Gicerin, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18, MUC18Gicerin | |
Rat | |
IgG2a | |
RUO | |
Primary | |
4162 | |
Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)