Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MME Mouse anti-Human, Unconjugated, Clone: MME/1870, Abnova
Mouse Monoclonal Antibody.
Manufacturer: Abnova Corporation MAB21309
Description
MME Monoclonal antibody specifically detects MME in Human samples. It is validated for IHC-P.Specifications
MME | |
MME/1870 | |
membrane metallo-endopeptidase | |
Liquid | |
Mouse | |
IgG2c, Kappa | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. | |
Monoclonal | |
Human |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
MME | |
CALLA, CD10, DKFZp686O16152, MGC126681, MGC126707, NEP | |
Recombinant protein corresponding to amino acids 297-483 of human MME | |
100 μg | |
Primary | |
4311 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title