Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMF, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
2756-87-8
C5H6O4
130.099
NKHAVTQWNUWKEO-NSCUHMNNSA-N
monomethyl fumarate,4-methoxy-4-oxobut-2-enoic acid,methyl hydrogen fumarate,mono-methyl fumarate,fumaric acid monomethyl ester,unii-45iub1px8r,monomethylfumarate,45iub1px8r,e-4-methoxy-4-oxobut-2-enoic acid,2e-4-methoxy-4-oxobut-2-enoic acid
5369209
(E)-4-methoxy-4-oxobut-2-enoic acid
COC(=O)C=CC(=O)O
This item is not returnable. View return policy
2756-87-8
C5H6O4
130.099
NKHAVTQWNUWKEO-NSCUHMNNSA-N
monomethyl fumarate,4-methoxy-4-oxobut-2-enoic acid,methyl hydrogen fumarate,mono-methyl fumarate,fumaric acid monomethyl ester,unii-45iub1px8r,monomethylfumarate,45iub1px8r,e-4-methoxy-4-oxobut-2-enoic acid,2e-4-methoxy-4-oxobut-2-enoic acid
5369209
(E)-4-methoxy-4-oxobut-2-enoic acid
COC(=O)C=CC(=O)O

Activator of Nrf2 pathway; primary metabolite of DMF (Cat. No. 4512)

Nuclear factor (erythroid-derived-2)-like 2 (Nrf2) pathway activator. Also exhibits agonist activity at GPR109A. Primary metabolite of DMF (Cat. No. 4512).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 130.1
Percent Purity >99%
Chemical Name or Material MMF

Recommended Storage : Store at Room Temperature