Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris™ MMG 11
TLR2 antagonist
$172.22 - $722.22
Specifications
Molecular Formula | C15H14O7 |
---|---|
Purity | 97% |
Target | Toll-like Receptor Antagonists |
Chemical Name or Material | Ethyl 5-(2-oxo-2-(2,3,4-trihydroxyphenyl)ethyl)furan-2-carboxylate |
CAS | 313254-94-3 |
Description
TLR2 antagonist (IC50 values are in the range 1.7 - 5.7 μM). Exhibits selectivity for TLR2 over TLR4, TLR5, TLR7/8 and TLR9. Reduces TLR2-mediated cytokine secretion from PBMCs.Specifications
C15H14O7 | |
Toll-like Receptor Antagonists | |
313254-94-3 |
97% | |
Ethyl 5-(2-oxo-2-(2,3,4-trihydroxyphenyl)ethyl)furan-2-carboxylate |
Safety and Handling
Recommended Storage : Store at -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title