Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMGT1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551190

Catalog No. PA551190


Add to cart

Description

Description

The antibody detects endogenous levels of total MMGT1 protein.

Mmgt1 encodes a protein that that mediates Mg(2+) transport.
Specifications

Specifications

MMGT1
Polyclonal
Unconjugated
MMGT1
Liquid
MMGT1
Full length fusion protein of human MMGT1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q8N4V1
EMC5, TMEM32
Rabbit
IgG
100 μL
-20°C
Primary
93380
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit