Learn More
GE Healthcare MMP-1 Human Biotrak ELISA System Available on GSA/VA Contract for Federal Government customers only.
Measure total human MMP-1 levels in one day. GE Healthcare MMP-1 Human Biotrak ELISA System is a ready-to-use kit featuring a coated 96-well microplate and all required buffers and standards. Biotrak ELISAs measure MMP protein levels in human serum, plasma and cell culture samples.
Manufacturer:  GE Healthcare RPN2610
Catalog No. 45-200-587
Add to cart
Description
Description
The MMP-1 Human Biotrak ELISA system has been specifically designed for research purposes. MMP-1 may be measured in the range 6.25-100ng/ml. The assay is based on a two site ELISA ‘sandwich’ format.
  • Measures total MMP-1
  • Specific for human MMP-1
  • Precise and accurate measurement
  • Non-isotopic protocol
  • Ready to use substrate
  • Same day protocol
Measurement protocol
  • Standards and samples are incubated in microplate wells pre-coated with anti-MMP-1 antibody
  • Any MMP-1 present will be bound to the wells, with other components of the sample being removed by washing and aspiration
  • A second polyclonal antibody to MMP-1 is then bound to the MMP-1; any excess is removed by washing and aspiration
  • The second antibody bound to the wells is detected using donkey anti-rabbit horseradish peroxidase
  • The amount of peroxidase bound to each well is determined by the addition of TMB ‘ready-to-use’ substrate
  • The reaction is stopped by addition of an acid solution and the resultant color read at 450nm in a microplate spectrophotometer
The concentration of MMP-1 in a sample is determined by interpolation from a standard curve. Each pack contains reagent for 96 determinations. This allows the construction of a standard curve plus the measurement of 42 samples in duplicate.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
6.25ng/mL to 100ng/mL
MMP-1, Human, Biotrak Assay.
Pipettes or pipetting equipment with disposable polypropylene tips (100 μl, 500 μl, 1 mL and 5 mL); Disposable polypropylene test tubes; Glass measuring cylinders (100 mL, and 1 l); Distilled or deionized water; Spectrophotometer plate reader capable of m
PP (Polypropylene)
1.7 ng/ml
-15 - -30°C
0.079
Suitable for use with serum, plasma (EDTA, heparin, citrate) and cell culture supernatant samples Biotrak ELISAs measure MMP protein levels in human serum, plasma, and cell culture samples
Microplate: 12 x 8 well strips coated with mouse anti-MMP-1; Assay buffer 1: 10mL of phosphate buffer concentrate; Assay buffer 2: 10mL of phosphate buffer concentrate; Standard: 1mL human MMP-1 (recombinant); Antibody: lyophilised rabbit anti-MMP-1; Peroxidase conjugate: lyophilised donkey anti-rabbit horseradish peroxidase; Wash buffer: 12.5mL phosphate buffer concentrate; TMB substrate: 3,3’,5,5’-Tetramethylbenzidine (TMB)/hydrogen peroxide
96 wells
The assay recognises total human MMP-1, ie. free MMP-1 and that complexed with inhibitors such as TIMP-1. It does not recognise MMP-1 bound by the non-specific protease inhibitor α2-macroglobulin. There is no cross-reactivity with α2-macroglobulin alone.
5.5-h