Learn More
MMP-1 Mouse anti-Human, Mouse, Alexa Fluor™ 350, Clone: 6A5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP222123AF350
Catalog No. NP222123A30
Add to cart
Description
Description
MMP-1 Monoclonal antibody specifically detects MMP-1 in Human,Mouse samples. It is validated for Western Blot,Flow Cytometry,ELISA,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin).
Specifications
Specifications
MMP-1
6A5
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
CLGmatrix metalloprotease 1, CLGN, EC 3.4.24, EC 3.4.24.7, Fibroblast collagenase, interstitial collagenase, matrix metallopeptidase 1 (interstitial collagenase), matrix metalloproteinase 1 (interstitial collagenase), Matrix metalloproteinase-1, MMP-1
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human, Mouse
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
MMP1
Purified recombinant fragment of human MMP1 expressed in E. coli. [UniProtNo. P03956]
0.1 mL
Primary
4312