Learn More
MMP-10 (Stromelysin-2) Ab-2 Mouse Monoclonal Antibody, Epredia™
Epredia™ MMP-10 (Stromelysin-2) Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.
Specifications
Antigen | MMP-10 (Stromelysin-2) Ab-2 |
---|---|
Clone | IVC5 |
Applications | Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Concentration | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Concentration | Quantity | Price | Quantity & Availability | ||||
MS822P0
|
Epredia
MS822P0 |
200μg/mL | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
MS822P1
|
Epredia
MS822P1 |
200μg/mL | 500 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
MS822P
|
Epredia
MS822P |
200μg/mL | 1 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
MS822P1ABX
|
Epredia
MS822P1ABX |
1mg/mL | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
MS822PABX
|
Epredia
MS822PABX |
1mg/mL | 200 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
Matrix metalloproteinases (MMP) are proteolytic enzymes capable of degrading extracellular matrix. Stromelysin, a member of the matrix metalloproteinase family, demonstrates wide substrate specificity with the ability to degrade proteoglycan, fibronectin, laminin, casein, and the nonhelical region of collagen. The two forms of stromelysin (SL), types 1 (MMP-3) and 2 (MMP-10), share 82% sequence homology, but exhibit differences in cellular synthesis and inducibility by cytokines and growth factors in vitro.Host Species: Mouse
Clone: IVC5
Isotype: IgG2b
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Human recombinant MMP-10 (stromelysin-2 protein)
Molecular Weight: ∽57kDa (proform) and ∽44kDa (active form)
Positive Control: Conditioned, serum-free medium from (TPA-treated) human large cell lymphoma U937 cells; Placenta; Bladder, breast and ovarian CAs
Cellular Localization: Cytoplasmic
Recommended for:
- Western Blotting
- Immunoprecipitation (Native verified)
Note:
USA: RUO; Int'l: RUO
Specifications
MMP-10 (Stromelysin-2) Ab-2 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Proteases And Inhibitors | |
Human recombinant MMP-10 (stromelysin-2 protein) | |
Primary |
IVC5 | |
Monoclonal | |
Concentrated | |
RUO | |
Human | |
IgG2b |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.