Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ MMP-11 (Stromelysin-3) Ab-5, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in experiments with the Thermo Scientific™ MMP-11 (Stromelysin-3) Ab-5, Mouse Monoclonal Antibody.

$237.50 - $878.00

Specifications

Antigen MMP-11 (Stromelysin-3) Ab-5
Clone SL3.05
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 10
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS1035P
View Documents
Thermo Scientific™
MS1035P
Purified Ab with BSA and Azide 1mL Each for $878.00
Add to cart
 
MS1035P0
View Documents
Thermo Scientific™
MS1035P0
Purified Ab with BSA and Azide 100μL Each for $237.50
Add to cart
 
MS1035P1
View Documents
Thermo Scientific™
MS1035P1
Purified Ab with BSA and Azide 500μL Each for $525.50
Add to cart
 
MS1035R7
View Documents
Thermo Scientific™
MS1035R7
Ready-to-use for IHC (UVLP) 7mL Each for $463.50
Add to cart
 
MS1035B
View Documents
Thermo Scientific™
MS1035B
Biotinylated Ab with BSA and Azide 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS1035R1
View Documents
Thermo Scientific™
MS1035R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
MS1035B0
View Documents
Thermo Scientific™
MS1035B0
Biotinylated Ab with BSA and Azide 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS1035P1ABX
View Documents
Thermo Scientific™
MS1035P1ABX
Ab without BSA/Azide (Frozen) 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS1035PABX
View Documents
Thermo Scientific™
MS1035PABX
Ab without BSA/Azide (Frozen) 200μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS1035B1
View Documents
Thermo Scientific™
MS1035B1
Biotinylated Ab with BSA and Azide 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in experiments with the Thermo Scientific™ MMP-11 (Stromelysin-3) Ab-5, Mouse Monoclonal Antibody.

Stromelysin, a member of the matrix metalloproteinase family, demonstrates wide substrate specificity with the ability to degrade proteoglycan, fibronectin, laminin, casein, and the nonhelical region of collagen. The stromelysin-3 (MMP-11) gene was originally identified on the basis that it is expressed specifically in stromal cells surrounding invasive breast carcinomas. MMP-11 is localized by in situ hybridization to the long arm of chromosome 22. The MMP-11 expression is found more specifically in malignant tumors than in benign ones.

Host Species: Mouse

Clone: SL3.05

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Immunogen: Recombinant protein corresponding to active form of human MMP-11

Molecular Weight: ~60kDa (latent), ~47kDa (active) and ~35kDa (degradation product)

Positive Control: Recombinant MMP-11 protein; Placenta

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native verified)
  • Affinity Purification (Not verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

MMP-11 (Stromelysin-3) Ab-5
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Murine
Proteases And Inhibitors
Recombinant protein corresponding to active form of human MMP-11
SL3.05
Monoclonal
RUO
Primary
IgG2a, κ
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit