Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ MMP-13 (Collagenase-3) Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MMP-13 (Collagenase-3) Ab-1, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ MS825P1

 View more versions of this product

Catalog No. MS825P1

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MMP-13 (Collagenase-3) Ab-1, Mouse Monoclonal Antibody.

Human collagenase-3 (MMP13) is a recently identified member of the matrix metalloproteinase (MMP) family that is expressed in breast carcinomas and in articular cartilage from arthritic patients. The MMP13 gene has been isolated and characterized. This gene is composed of 10 exons and 9 introns and spans over 12.5 kb. The overall organization of the collagenase-3 gene is similar to that of other MMP genes clustered at chromosome 11q22.

Host Species: Mouse

Clone: VIIIA2

Isotype: IgG1/κ

Species Reactivity: Human, Mouse and Rabbit. Others not known.

Epitope: Not determined

Immunogen: Human recombinant collagenase-3 protein

Molecular Weight: ~60kDa (proform) and ~48kDa (active form)

Positive Control: Conditioned, serum-free medium from (TPA-treated) human chondrosarcoma SW1353 cells; Placenta; Bladder, breast and ovarian CAs

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunoprecipitation (Native verified)
  • Affinity Purification (Not verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

MMP-13 (Collagenase-3) Ab-1
Monoclonal
200μg/mL
Purified Ab with BSA and Azide
Murine
IgG2a kappa
RUO
Primary
Monoclonal
Affinity Purification, Immunohistochemistry (Paraffin), Immunoprecipitation
VIIIA2
Unconjugated
Concentrated
Human recombinant collagenase-3 protein
500μL
Proteases And Inhibitors
Primary
Human, Murine, Rabbit
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit