Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-13 Mouse anti-Human, Mouse, Rat, DyLight 680, Clone: OTI2D8, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272740FR
Description
MMP-13 Monoclonal specifically detects MMP-13 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockdown Validated.Specifications
MMP-13 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
MMP13 | |
Recombinant protein fragment corresponding to amino acids 104-471 of human MMP-13 (NP_002418) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, KnockDown | |
OTI2D8 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, KnockDown Validated | |
CLG 3, CLG3EC 3.4.24, collagenase 3, EC 3.4.24.-, EC 3.4.24.22, EC 3.4.24.24, EC 3.4.24.35, EC 3.4.24.65, EC 3.4.24.7, MANDP1, matrix metallopeptidase 13 (collagenase 3), matrix metalloproteinase 13 (collagenase 3), Matrix metalloproteinase-13, MMP-13 | |
Mouse | |
Affinity Purified | |
Cancer, Cellular Markers, Extracellular Matrix | |
4322.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title