Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ MMP-14/MT1-MMP Ab-2, Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in experiments with the Thermo Scientific™ MMP-14/MT1-MMP Ab-2, Rabbit Polyclonal Antibody.

Specifications

Antigen MMP-14/MT1-MMP Ab-2
Concentration 1mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Concentration Quantity Format Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Quantity Format Price Quantity & Availability  
RB1545P1
View Documents
Thermo Scientific™
RB1545P1
1mg/mL 500μL Purified with BSA and azide This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in experiments with the Thermo Scientific™ MMP-14/MT1-MMP Ab-2, Rabbit Polyclonal Antibody.

MMPs are a group of enzymes involved in matrix degradation. Of the sixteen members of matrix metalloproteinase family ten exist in soluble form whereas MT-MMPs exist as integral membrane proteins. MT1-MMP, MT2-MMP, MT3-MMP also known as MMP14, MMP15, MMP16 respectively contain a C-terminal transmembrane domain anchoring them to the cell surface. They have an 8 amino acid insert in their catalytic domain. MT1-MMP cleaves progelatinase A (MMP-2) and progelatinase B to their active forms. MT1-MMP gets activated through the membrane plasmin cascade. It binds to TIMP-2 first and then to MMP-2 forming a trimer. It cleaves the pro-MMP-2 to its active form.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Others not known.

Epitope: Hemopexin-like domain

Immunogen: A synthetic peptide from the hemopexin-like domain of human MMP-14/MT1-MMP

Molecular Weight: ~66kDa (latent) and ~54kDa (active)

Positive Control: Recombinant MMP-14 protein; Conditioned, serum-free medium from (TPA-treated ) HFL-1 cells

Cellular Localization: Membrane and cytoplasmic

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

MMP-14/MT1-MMP Ab-2
Western Blot
Unconjugated
RUO
Primary
1mg/mL
Polyclonal
Rabbit
Proteases And Inhibitors
IgG
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit