Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-14/MT1-MMP Antibody (3G4.2) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$243.50 - $792.00

Specifications

Antigen MMP-14/MT1-MMP
Clone 3G4.2
Concentration 1 mg/mL
Dilution Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB019446
SDP
View Documents
Novus Biologicals
NBP2297350.025MG
0.025 mg
Each for $243.50
Only null left
Add to Cart
 
NBP229735
SDP
View Documents
Novus Biologicals
NBP229735
0.1 mg
Each for $792.00
Only null left
Add to Cart
 
Description

Description

MMP-14/MT1-MMP Monoclonal specifically detects MMP-14/MT1-MMP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

MMP-14/MT1-MMP
1 mg/mL
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Unconjugated
Mouse
Cancer, Extracellular Matrix
P50281
4323
Recombinant protein containing the catalytic domain of human MT1-MMP.
Primary
Store at 4C. Do not freeze.
60 kDa
3G4.2
Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500
Monoclonal
Purified
RUO
Human
EC 3.4.24, EC 3.4.24.80, matrix metallopeptidase 14 (membrane-inserted), matrix metalloproteinase 14 (membrane-inserted), matrix metalloproteinase-14, membrane type 1 metalloprotease, Membrane-type matrix metalloproteinase 1, Membrane-type-1 matrix metalloproteinase, MMP-14, MMP-X1, MT1MMP, MT1-MMPMTMMP1, MT-MMP 1
MMP14
IgG2b κ
Protein A purified
Clone 3G4.2 recognizes the catalytic domain of MT1-MMP, identifying the active form of the protein. MW is approximately 60 kDa.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.