Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-14/MT1-MMP Mouse anti-Human, Clone: 114-6G6, Novus Biologicals™
Mouse Monoclonal Antibody
$212.55 - $723.00
Specifications
Antigen | MMP-14/MT1-MMP |
---|---|
Clone | 114-6G6 |
Concentration | 2 mg/mL |
Dilution | Western Blot 8-20 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Western Blot, ELISA, Immunohistochemistry (Paraffin) |
Description
MMP-14/MT1-MMP Monoclonal specifically detects MMP-14/MT1-MMP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MMP-14/MT1-MMP | |
2 mg/mL | |
Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
EC 3.4.24, EC 3.4.24.80, matrix metallopeptidase 14 (membrane-inserted), matrix metalloproteinase 14 (membrane-inserted), matrix metalloproteinase-14, membrane type 1 metalloprotease, Membrane-type matrix metalloproteinase 1, Membrane-type-1 matrix metalloproteinase, MMP-14, MMP-X1, MT1MMP, MT1-MMPMTMMP1, MT-MMP 1 | |
MMP14 | |
IgG1 κ | |
Affinity Purified | |
The antibody specifically reacts with human MT1-MMP. This is a purified mouse monoclonal antibody to an oligopeptide of REVPYAYIREGHEK (residue 160-173) on human membrane-type 1 matrix metalloproteinase (human MT1-MMP) and does not react with mouse or rat. However, this sequence is identical in mouse and rat MT1-MMP. For unknown reasons this antibody does not react on westerns to CC1043. |
114-6G6 | |
Western Blot 8-20 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry-Paraffin 1:10-1:500 | |
Monoclonal | |
Mouse | |
Cancer, Extracellular Matrix | |
P50281 | |
4323 | |
This antibody is specific to MMP-14 | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title