Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-17 Mouse anti-Human, Alexa Fluor™ 405, Clone: MM0638-12N24, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232725AF405

Catalog No. NP23272A40


Add to cart

Description

Description

MMP-17 Monoclonal antibody specifically detects MMP-17 in Human samples. It is validated for ELISA.
Specifications

Specifications

MMP-17
MM0638-12N24
ELISA
Conjugated, Purified
MMP17
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with recombinant human MMP-17
Protein G purified
Store at 4°C in the dark.
Monoclonal
Human
ELISA
Alexa Fluor™ 405
50 mM sodium borate with 0.05% sodium azide
matrix metallopeptidase 17 (membrane-inserted), matrix metalloproteinase 17 (membrane-inserted), matrix metalloproteinase-17, membrane-type matrix metalloproteinase 4, membrane-type-4 matrix metalloproteinase, MMP-17, MT4MMP, MT4-MMP, MT-MMP 4, MTMMP4
Mouse
IgG2
0.1 mL
Primary
4326.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit