Learn More
MMP-17 Mouse anti-Human, DyLight 405, Clone: MM0638-12N24, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232725V
Catalog No. NBP232725V
Add to cart
Description
Description
MMP-17 Monoclonal antibody specifically detects MMP-17 in Human samples. It is validated for ELISA.
Specifications
Specifications
MMP-17
MM0638-12N24
ELISA
Purified
MMP17
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with recombinant human MMP-17
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
ELISA
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
matrix metallopeptidase 17 (membrane-inserted), matrix metalloproteinase 17 (membrane-inserted), matrix metalloproteinase-17, membrane-type matrix metalloproteinase 4, membrane-type-4 matrix metalloproteinase, MMP-17, MT4MMP, MT4-MMP, MT-MMP 4, MTMMP4
Mouse
IgG2
0.1 ml
Primary
4326