Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ MMP-2 (72kDa Collagenase IV) Ab-2, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in experiments with the Thermo Scientific™ MMP-2 (72kDa Collagenase IV) Ab-2, Mouse Monoclonal Antibody.

$174.40 - $1014.00

Specifications

Antigen MMP-2 (72kDa Collagenase IV) Ab-2
Clone VB3
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Format Concentrated
View More Specs

Products 8
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity & Availability  
MS804B
View Documents
Thermo Scientific™
MS804B
200μg/mL Biotin 1mL
Each for $1,014.00
Add to cart
 
MS804B0
View Documents
Thermo Scientific™
MS804B0
200μg/mL Biotin 100μL
Each for $233.50
Add to cart
 
MS804B1
View Documents
Thermo Scientific™
MS804B1
200μg/mL Biotin 500μL
Each for $633.50
Add to cart
 
MS804P
View Documents
Thermo Scientific™
MS804P
200μg/mL Unconjugated 1mL
Each for $807.00
Add to cart
 
MS804P0
View Documents
Thermo Scientific™
MS804P0
200μg/mL Unconjugated 100μL
Each for $174.40
Add to cart
 
MS804P1
View Documents
Thermo Scientific™
MS804P1
200μg/mL Unconjugated 500μL
Each for $482.00
Add to cart
 
MS804P1ABX
View Documents
Thermo Scientific™
MS804P1ABX
1mg/mL Unconjugated 100μL
Each for $589.00
Add to cart
 
MS804PABX
View Documents
Thermo Scientific™
MS804PABX
1mg/mL Unconjugated 200μL
Each for $916.00
Add to cart
 
Description

Description

Ensure accurate, reproducible results in experiments with the Thermo Scientific™ MMP-2 (72kDa Collagenase IV) Ab-2, Mouse Monoclonal Antibody.

MMPs are proteolytic enzymes capable of degrading connective tissue components. They have a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and are inhibited by specific tissue inhibitors of metalloproteinases (TIMPs). MMPs and TIMPs play a significant role in regulating angiogenesis. MMP-2; also known as 72kDa collagenase IV or gelatinase A is synthesized as a 631 amino acid proenzyme which is activated by cleavage of the first 80 amino acids.

Host Species: Mouse

Clone: VB3

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human native 72kDa MMP-2

Molecular Weight: 72kDa (proform) and ~66kDa (active form)

Positive Control: Conditioned, serum-free medium from (TPA-treated) HFL-1 cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Not Suitable)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

MMP-2 (72kDa Collagenase IV) Ab-2
Immunofluorescence, Immunoprecipitation, Western Blot
Concentrated
RUO
Primary
IgG2a kappa
VB3
Monoclonal
Murine
Proteases And Inhibitors
Human native 72kDa MMP-2
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit