Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-2 Mouse anti-Human, Mouse, Rat, Guinea Pig, PE-Cyanine7, Clone: 2C1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NB200113PECY7

Catalog No. N20113PECY7

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

MMP-2 Monoclonal antibody specifically detects MMP-2 in Human, Mouse, Rat, Guinea Pig samples. It is validated for Flow Cytometry.
Specifications

Specifications

MMP-2
2C1
Flow Cytometry
Conjugated, Purified
MMP2
Activated recombinant human MMP2. [UniProt No. P08253]
Protein G purified
Angiogenesis, Apoptosis, Cancer, Cell Biology, Cytoskeleton Markers, Extracellular Matrix, HIF Target Genes, Hypoxia, Metastasis, Tumor Suppressors
Primary
4313.0
Guinea Pig, Human, Mouse, Rat
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
72 kDa gelatinase, CLG4, CLG4A72 kDa type IV collagenase, collagenase type IV-A, EC 3.4.24, EC 3.4.24.24, Gelatinase A, matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase), matrix metalloproteinase 2 (gelatinase A, 72kD gelatinase, 72kD type IVcollagenase), Matrix metalloproteinase-2, matrix metalloproteinase-II, MMP-2, MMP-II, MONA, neutrophil gelatinase, TBE-1matrix metalloproteinase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase)
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
This is specific for pro and active MMP-2.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit