Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-3, Mouse anti-Human, DyLight 550, Clone: MMP3/2655, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279865R
Description
MMP-3 Monoclonal antibody specifically detects MMP-3 in Human samples. It is validated for Western Blot, ELISA.Specifications
MMP-3 | |
MMP3/2655 | |
Western Blot, ELISA | |
Conjugated, Purified | |
MMP3 | |
Recombinant fragment of human MMP-3 protein (around aa 236-363) (exact sequence is proprietary) | |
Protein A purified | |
RUO | |
Store at 4°C in the dark. | |
Monoclonal |
ELISA, Western Blot | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CHDS6, EC 3.4.24, EC 3.4.24.17, matrix metallopeptidase 3 (stromelysin 1, progelatinase), matrix metalloproteinase 3 (stromelysin 1, progelatinase), Matrix metalloproteinase-3, MGC126102, MGC126103, MMP-3, proteoglycanase, SL-1, STMY, STMY1MGC126104, STR1, stromelysin-1, transin-1 | |
Mouse | |
IgG2b Kappa | |
0.1 mL | |
Angiogenesis, Cancer, Extracellular Matrix | |
Primary | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title