Learn More
MMP-3 (Stromelysin-1) Ab-1 Mouse Monoclonal Antibody, Epredia™
Epredia™ MMP-3 (Stromelysin-1) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
$815.00
Specifications
Antigen | MMP-3 (Stromelysin-1) Ab-1 |
---|---|
Clone | SL-1 ID3 |
Applications | Immunofluorescence, Western Blot |
Classification | Monoclonal |
Form | Concentrated |
Catalog Number | Mfr. No. | Conjugate | Concentration | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Conjugate | Concentration | Quantity | Price | Quantity & Availability | |||
MS809B1
|
Epredia
MS809B1 |
Biotin | 200μg/mL | 500 μL |
Each for $815.00
|
|
|||
MS809B0
|
Epredia
MS809B0 |
Biotin | 200μg/mL | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS809B
|
Epredia
MS809B |
Biotin | 200μg/mL | 1 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS809P0
|
Epredia
MS809P0 |
Unconjugated | 200μg/mL | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS809P1
|
Epredia
MS809P1 |
Unconjugated | 200μg/mL | 500 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS809P
|
Epredia
MS809P |
Unconjugated | 200μg/mL | 1 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS809P1ABX
|
Epredia
MS809P1ABX |
Unconjugated | 1mg/mL | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS809PABX
|
Epredia
MS809PABX |
Unconjugated | 1mg/mL | 200 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
Description
Matrix metalloproteinases (MMP) are proteolytic enzymes capable of degrading connective tissue components. Imbalanced secretion of certain MMP or disturbances in the differential control of MMP by tissue inhibitor of MMPs (TIMPs) are implicated in the invasive potential of malignant tumors. Stromelysin (MMP3/Transin) degrades several matrix components including the core proteins of proteoglycans, laminin and non-helical regions of collagens.Host Species: Mouse
Clone: SL-1 ID3
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: APMA (4-Aminophenylmercuric acetate) activated Human stromelysin-1 (SL-1)
Molecular Weight: 54/59kDa (proform) and 44/49kDa (active form)
Positive Control: Conditioned, serum-free medium from TPA-treated human endometrial cells
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Not Suitable)
- Western Blotting
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
General References:1. Lein M, et al. Urologe A 199837(4):377-381.
2. Clavel C, et al. Bull Cancer 199279:261-270.
3. Muller JC, et al. Biol Chem 1997378:1475-1480.
Specifications
MMP-3 (Stromelysin-1) Ab-1 | |
Immunofluorescence, Western Blot | |
Concentrated | |
RUO | |
Human | |
IgG2a κ |
SL-1 ID3 | |
Monoclonal | |
Mouse | |
Proteases And Inhibitors | |
APMA (4-Aminophenylmercuric acetate) activated Human stromelysin-1 (SL-1) | |
Primary |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.