Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-8 Mouse anti-Human, DyLight 755, Clone: MM0023-7A11, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11060989IR
Description
MMP-8 Monoclonal specifically detects MMP-8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MMP-8 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MMP8 | |
Human recombinant MMP-8 | |
0.1 mL | |
Primary | |
This antibody recognizes human MMP-8 in western blot without cross reactivity with other MMPs. | |
Store at 4C in the dark. | |
IgG2 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0023-7A11 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CLG1HNC, EC 3.4.24, EC 3.4.24.34, matrix metallopeptidase 8 (neutrophil collagenase), matrix metalloproteinase 8 (neutrophil collagenase), Matrix metalloproteinase-8, MMP-8, neutrophil collagenase, PMNL collagenase, PMNL-CL | |
Mouse | |
Protein G purified | |
Cancer | |
4317 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title