Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-9 Mouse anti-Human, Mouse, Rat, Alexa Fluor 750, Clone: 4A3, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP213173AF750
Description
MMP-9 Monoclonal specifically detects MMP-9 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MMP-9 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
MMP9 | |
Synthetic peptide from the N-terminus of human MMP9 [Swiss-ProtNo. P14780] | |
0.1 mL | |
Primary | |
Active MMP9. It does not react with the MMP9 proenzyme or the active and proenzyme forms of MMP2. | |
Store at 4°C in the dark. |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
4A3 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase | |
Mouse | |
Protein G purified | |
Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, GPCR | |
4318.0 | |
Human, Mouse, Rat | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title