Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-9 Rabbit anti-Human, Clone: 31, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | MMP-9 |
---|---|
Clone | 31 |
Dilution | ELISA 1:1000-1:2000 |
Applications | ELISA |
Classification | Monoclonal |
Description
MMP-9 Monoclonal specifically detects MMP-9 in Human samples. It is validated for ELISA.Specifications
MMP-9 | |
ELISA 1:1000-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase | |
Recombinant Human MMP-9 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
31 | |
ELISA | |
Unconjugated | |
Rabbit | |
Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, GPCR | |
0.2 um filtered solution in PBS | |
4318 | |
IgG | |
Protein A purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title