Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-9 Rabbit anti-Human, DyLight 350, Clone: 43, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289415UV
Description
MMP-9 Monoclonal specifically detects MMP-9 in Human samples. It is validated for Sandwich ELISA Detection.Specifications
MMP-9 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
43 | |
Sandwich ELISA Detection | |
92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human MMP-9 (NP_004985.2; Met1-Asp707). | |
0.1 mL | |
Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, GPCR | |
4318 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title