Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-9 Rabbit anti-Human, Guinea Pig, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
$236.16 - $781.00
Specifications
Antigen | MMP-9 |
---|---|
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB6001217
|
Novus Biologicals
NB6001217 |
0.5 mL |
Each for $781.00
|
|
NB600121725
|
Novus Biologicals
NB60012170.25ML |
0.25 mL |
Each for $436.00
|
|
NB018206
|
Novus Biologicals
NB60012170.1ML |
0.1 mL |
Each for $236.16
|
|
Description
MMP-9 Polyclonal specifically detects MMP-9 in Human, Guinea Pig, Bovine (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
MMP-9 | |
Polyclonal | |
Rabbit | |
4318 | |
IgG | |
This reacts with the 92 kDa pro form and 86 kDa active form of MMP-9. |
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase | |
MMP9 | |
Primary |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title