Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-9 Rabbit anti-Mouse, Alexa Fluor 594, Clone: 2, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP290636AF594

Catalog No. NB040408


Add to cart

Description

Description

MMP-9 Monoclonal antibody specifically detects MMP-9 in Mouse samples. It is validated for Western Blot
Specifications

Specifications

MMP-9
2
Western Blot
Purified
Rabbit
IgG
0.1 mL
Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, GPCR
Primary
4318
Western Blot
Alexa Fluor 594
50mM Sodium Borate
92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase
Recombinant Mouse MMP-9 Protein
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit