Learn More
Alfa Aesar™ MMP Inhibitor II
A potent, reversible, broad range inhibitor of matrix metalloproteinases
Manufacturer:  Alfa Aesar™ J65638MA
 View more versions of this product
Catalog No. AAJ65638MA
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
10mg
−30° to −10°C
A potent, reversible, broad range inhibitor of matrix metalloproteinases
C21H27N3O8S2
N-Hydroxy-1,3-bis(4-methoxyphenylsulfonyl)-5,5-dimethylhexahydropyrimidine-2-carboxamide
Signal Transduction Reagent
Off-white
Solid
203915-59-7
513.58g/mol
Soluble in DMSO