Learn More
Aviva Systems Biology Corp MMP10 Antibody - N-terminal region (AVARP09017_T100) | 100uL
Supplier:  Aviva Systems Biology Corp AVARP09017_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MMP10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MMP10. Alternate names: SL-2, STMY2ApplicationWB. Species reactivity: Human
Catalog No. 50-014-62570