Learn More
Invitrogen™ MMP12 Recombinant Rabbit Monoclonal Antibody (1O3A4)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542836
Description
Positive test controls include: A-549, BxPC-3, Mouse lung, Mouse brain, Mouse spleen, Rat lung. The target is usually found in the following locations: Secreted, extracellular matrix, extracellular space. Immunogen sequence: FGFPNFVKKI DAAVFNPRFY RTYFFVDNQY WRYDERRQMM DPGYPKLITK NFQGIGPKID AVFYSKNKYY YFFQGSNQFE YDFLLQRITK TLKSNSWFGC.
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. It is thought that the protein encoded by this gene is cleaved at both ends to yield the active enzyme, but this processing has not been fully described. The enzyme degrades soluble and insoluble elastin. It may play a role in aneurysm formation and studies in mice suggest a role in the development of emphysema. The gene is part of a cluster of MMP genes which localize to chromosome 11q22. 3.
Specifications
MMP12 | |
Recombinant Monoclonal | |
0.5 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P34960, P39900, Q63341 | |
Mmp12 | |
A synthetic peptide corresponding to a sequence within amino acids 371-470 of human MMP12 (P39900). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
1O3A4 | |
Unconjugated | |
Mmp12 | |
3.4.24.65; AV378681; HME; Macrophage elastase; macrophage metalloelastase; macrophage-metalloelastase; matrix metallo protease; matrix metallopeptidase 12; matrix metallopeptidase 12 (macrophage elastase); matrix metalloproteinase 12; matrix metalloproteinase 12 (macrophage elastase); matrix metalloproteinase-12; ME; MGC138506; MME; Mmel; MMP; Mmp12; MMP-12; MMPs | |
Rabbit | |
Affinity Chromatography | |
RUO | |
117033, 17381, 4321 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.