Learn More
Abnova™ MMP13 (Human) ELISA Kit
Sandwich enzyme-linked immunosorbent assay
Manufacturer:  Abnova™ KA5682
Catalog No. 89-339-613
Add to cart
Description
Description
MMP13 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for the quantitative measurement of human MMP13 in cell culture supernates, serum and plasma (heparin).
Specifications
Specifications
Quantitative
MMP13
1 Kit
RUO
Human
4322
Standard curve
Store at 4°C for 6 months, at -20°C for 12 months. Avoid multiple freeze-thaw cycles.
CLG3