Learn More
Abnova™ MMP14 (Human) ELISA Kit
Use to measure quantitatively human MMP14. Sandwich enzyme immunoassay.
Manufacturer:  Abnova™ KA5078
Catalog No. 89-334-062
Add to cart
Description
Description
MMP14 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for the quantitative measurement of human MMP14.
Specifications
Specifications
P50281
4323
Store at 4°C for 6 months, at -20°C for 12 months. Avoid multiple freeze-thaw cycles.
Human
Quant
ELISA Kit
1 Kit