Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP14 Mouse anti-Human, Clone: 133CT15.10.5.1, Abnova™

Mouse monoclonal antibody raised against synthetic peptide of human MMP14.

Manufacturer:  Abnova Corporation MAB12762

Catalog No. 89-934-563


Add to cart

Description

Description

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. However, the protein encoded by this gene is a member of the membrane-type MMP (MT-MMP) subfamily; each member of this subfamily contains a potential transmembrane domain suggesting that these proteins are expressed at the cell surface rather than secreted. This protein activates MMP2 protein, and this activity may be involved in tumor invasion. [provided by RefSeq]

Specifications

Specifications

MMP14
133CT15.10.5.1
Mouse monoclonal antibody raised against synthetic peptide of human MMP14.
In PBS (0.09% sodium azide).
MMP-X1/MT1-MMP/MTMMP1
Mouse
IgM
400 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunofluorescence, Western Blot
Unconjugated
Immunofluorescence (1:10 - 1:50) Western Blot (1:100 - 1:500) The optimal working dilution should be determined by the end user.
Liquid
MMP14
A synthetic peptide (conjugated with KLH) corresponding to amino acids 470-499 at C-terminus region of human MMP14.
Euglobin precipitation
RUO
Primary
4323
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit