Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MMP19 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534748
Description
Recommended positive controls: human synovial fluid, MMP19-transfected 293T. Predicted reactivity: Rat (81%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MMP-19 (RASI-1, MMP-18) is a 508 amino acid peptide, originally isolated as an autoantigen from the synovium of a patient suffering from rheumatoid arthritis (RA). Its presence on the surface of activated PBMCs, TH1 lymphocytes, and Jurkat T lymphoma cells in the synovium of acute RA patients, suggests that MMP-19 plays a role in RA-associated joint tissue destruction. MMP-19 exists in the smooth muscle cells of the tunica media of large blood vessels of the RA patient, but not in the endothelial cell layer. Acutely inflamed tissue synovial capillaries strongly express MMP-19 in the cytoplasm. Regulated induction of MMP-19 in capillary endothelial cells during acute inflammation suggest a role in angiogenesis. MMP-19 is a single 2.7kb transcript found in mammary gland, placenta, lung, pancreas, ovary, small intestine, spleen, thymus, prostate, testis, colon, and heart.
Specifications
MMP19 | |
Polyclonal | |
Unconjugated | |
MMP19 | |
CODA; matrix metallo protease; matrix metallopeptidase 19; matrix metalloproteinase 19; matrix metalloproteinase RASI; Matrix metalloproteinase-18; matrix metalloproteinase-19; matrix metalloproteinase-beta19; MMP; MMP18; MMP-18; Mmp19; MMP-19; MMPs; Rasi; RASI-1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4327 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q99542 | |
MMP19 | |
Recombinant fragment corresponding to a region within amino acids 210 and 508 of MMP19 (Uniprot ID#Q99542). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction