Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP2 Mouse anti-Human, Clone: MMP2/1501, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14594

Catalog No. 89-034-677


Add to cart

Description

Description

Mouse monoclonal antibody raised against partial recombinant human MMP2.

  • High specificity
  • High reactivity
Specifications

Specifications

MMP2
MMP2/1501
Mouse monoclonal antibody raised against partial recombinant human MMP2.
In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Liquid
MMP2
Recombinant protein corresponding to amino acids 444-575 of human MMP2.
Protein A/G purification
RUO
Primary
4313
Flow Cytometry, Immunofluorescence, Western Blot
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
P08253
CLG4|CLG4A|MMP-II|MONA|TBE-1
Mouse
IgG1
100 ug
Store at 4°C.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)