Learn More
MMP2 Rabbit anti-Human, Mouse, Rat, Unconjugated, Clone: ADGF-13, Abnova™
Rabbit Monoclonal Antibody.
Manufacturer:  Abnova Corporation MAB22013
Catalog No. 89-339-751
Add to cart
Description
Description
MMP2 Monoclonal antibody specifically detects MMP2 in Human, Mouse, Rat samples. It is validated for ICC, WB-Ce, IF.
Specifications
Specifications
MMP2
ADGF-13
matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase)
Liquid
Rabbit
IgG
Store at -20°C for one year. It can also be aliquotted and stored frozen at -20°C for a longer time. Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human, Mouse, Rat
Immunocytochemistry, Western Blot, Immunofluorescence
Unconjugated
MMP2
CLG4, CLG4A, MMP-II, MONA, TBE-1
A synthetic peptide corresponding to human MMP2
100 μg
Primary
4313