Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MMP21 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5102884
Description
Antibody detects endogenous levels of total MMP21.
This gene encodes a member of the matrix metalloproteinase family. Proteins in this family are involved in the breakdown of extracellular matrix for both normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, and disease processes, such as asthma and metastasis. The encoded protein may play an important role in embryogenesis, particularly in neuronal cells, as well as in lymphocyte development and survival.
Specifications
MMP21 | |
Polyclonal | |
Unconjugated | |
MMP21 | |
b2b2458Clo; b2b873Clo; HTX7; matrix metallo protease; matrix metallopeptidase 21; matrix metallopeptidase 21 L homeolog; matrix metalloproteinase 21; matrix metalloproteinase-21; mmp; mmp21; MMP-21; mmp21.L; mmp21-A; MMPs; XELAEV_18034590mg; XMMP | |
Rabbit | |
Affinity chromatography | |
RUO | |
118856, 214766 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8K3F2, Q8N119 | |
MMP21 | |
A synthesized peptide derived from human MMP21(Accession Q8N119), corresponding to amino acid residues Y513-H563. | |
100 μL | |
Primary | |
Human, Mouse, Monkey | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction