Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP23 Polyclonal Antibody
MMP23 Polyclonal Antibody
Supplier: Thermo Scientific PA599177
Description
MMP23 Polyclonal Antibody for Western Blot, ICC/IF

Specifications
MMP23 | |
Polyclonal | |
Unconjugated | |
MMP23B | |
Femalysin; matrix metallo protease; matrix metallopeptidase 23A (pseudogene); matrix metallopeptidase 23B; matrix metalloproteinase 22; matrix metalloproteinase 23B; matrix metalloproteinase in the female reproductive tract; matrix metalloproteinase-21; Matrix metalloproteinase-22; matrix metalloproteinase-23; Matrix metalloproteinase-23, soluble form; MIFR; MIFR-1; MMP; MMP 23; MMP21; MMP-21; MMP22; MMP-22; MMP23; MMP-23; MMP23A; MMP23B; MMPs; RP11-345P4.2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
8510, 8511 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O75900 | |
MMP23B | |
A synthesized peptide derived from human MMP23A; MMP23B(Accession O75900), corresponding to amino acid residues L351-G390. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction