Learn More
Invitrogen™ MMP23 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113829
Description
Immunogen sequence: QKILHKKGKV YWYKDQEPLE FSYPGYLALG EAHLSIIANA VNEGTYTCVV For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 79%.
MMP23B (matrix metallopeptidase 23B) is also named as MMP21, MMP22. It is a regulator in matrix remodeling and may be playing a role in enchondral bone formation and in regulation of cranial suture closure. It has 3 isoforms produced by alternative splicing with the molecular weight of 44 kDa, 48 kDa and 46 kDa, respectively. It also can be N-glycosylated.
Specifications
MMP23 | |
Polyclonal | |
Unconjugated | |
MMP23B | |
Femalysin; matrix metallo protease; matrix metallopeptidase 23A (pseudogene); matrix metallopeptidase 23B; matrix metalloproteinase 22; matrix metalloproteinase 23B; matrix metalloproteinase in the female reproductive tract; matrix metalloproteinase-21; Matrix metalloproteinase-22; matrix metalloproteinase-23; Matrix metalloproteinase-23, soluble form; MIFR; MIFR-1; MMP; MMP 23; MMP21; MMP-21; MMP22; MMP-22; MMP23; MMP-23; MMP23A; MMP23B; MMPs; RP11-345P4.2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
8510, 8511 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O75900 | |
MMP23B | |
Synthetic peptide directed towards the middle region of human MMP23B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.