Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP23B Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP160036
Description
MMP23B Polyclonal specifically detects MMP23B in Human samples. It is validated for Western Blot.Specifications
MMP23B | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
O75900 | |
MMP23B | |
Synthetic peptides corresponding to MMP23B(matrix metallopeptidase 23B) The peptide sequence was selected from the middle region of MMP23B. Peptide sequence QKILHKKGKVYWYKDQEPLEFSYPGYLALGEAHLSIIANAVNEGTYTCVV. | |
100 μL | |
Signal Transduction | |
8510 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
EC 3.4.24.-, Femalysin, matrix metallopeptidase 23B, matrix metalloproteinase 22, matrix metalloproteinase 23B, matrix metalloproteinase in the female reproductive tract, Matrix metalloproteinase-21, Matrix metalloproteinase-22, matrix metalloproteinase-23, MIFR, MIFR-1MMP22femalysin, MMP21, MMP-21, MMP-22, MMP-23, MMP23A | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Guinea pig: 100%; Human: 100%; Chicken: 78%. | |
Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Zebrafish | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title