Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP24 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA551333
Description
The antibody detects endogenous levels of total MMP24 protein.
MMP24 Polyclonal antibody specifically detects MMP24 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
MMP24 | |
Polyclonal | |
Unconjugated | |
MMP24 | |
AU040325; matrix metallo protease; matrix metallopeptidase 24; matrix metallopeptidase 24 (membrane-inserted); matrix metalloproteinase 24; matrix metalloproteinase 24 (membrane-inserted); matrix metalloproteinase-21; Matrix metalloproteinase-24; membrane type 5-MMP; membrane-type 5 matrix metalloproteinase; membrane-type matrix metalloproteinase 5; Membrane-type-5 matrix metalloproteinase; MMP; Mmp21; MMP-21; Mmp24; MMP-24; MMP25; MMPs; MT5MMP; Mt5-mmp; MT-MMP 5; MTMMP5; MT-MMP5; Processed matrix metalloproteinase-24; RP4-614O4.10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10893, 17391, 83513 | |
-20°C | |
Liquid |
Western Blot | |
1.3 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q99PW6, Q9R0S2, Q9Y5R2 | |
MMP24 | |
Synthetic peptide of human MMP24 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction